Merck
  • Loss of G2A promotes macrophage accumulation in atherosclerotic lesions of low density lipoprotein receptor-deficient mice.
Loss of G2A promotes macrophage accumulation in atherosclerotic lesions of low density lipoprotein receptor-deficient mice.
Journal of lipid research (2005-04-19)
Brian W Parks, Ginger P Gambill, Aldons J Lusis, Janusz H S Kabarowski
ABSTRACT
Lysophosphatidylcholine (LPC) is considered a major proatherogenic component of oxidized low density lipoprotein based on its proinflammatory actions in vitro. LPC stimulates macrophage and T-cell chemotaxis via the G protein-coupled receptor G2A and may thus promote inflammatory cell infiltration during atherosclerotic lesion development. However, G2A also mediates proapoptotic effects of LPC and may therefore promote the death of inflammatory cells within developing lesions. To determine how these effects of LPC modify atherogenesis, we examined atherosclerotic lesion development in G2A-sufficient and G2A-deficient low density lipoprotein receptor knockout mice. Although LPC species capable of activating G2A-dependent responses were increased during lesion development, G2A-deficient mice developed lesions similar in size to those in their G2A-sufficient counterparts. Loss of G2A during atherosclerotic lesion development did not reduce macrophage and T-cell infiltration but instead resulted in increased lesional macrophage content associated with reduced numbers of terminal deoxynucleotidyl transferase-mediated dUTP nick end labeled cells and decreased collagen deposition. These data indicate that the ability of LPC to stimulate macrophage and T-cell chemotaxis via G2A is not manifested in vivo and that G2A-mediated proapoptotic rather than chemotactic action is most penetrant during atherogenesis and may modify the stability of atherosclerotic lesions by promoting macrophage death.
Sigma-Aldrich
ApopTag Fluorescein In Situ Apoptosis Detection Kit, The ApopTag Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a Fluorescein reporter molecule.