Merck
  • Minimal Ras-binding domain of Raf1 can be used as an activation-specific probe for Ras.
Minimal Ras-binding domain of Raf1 can be used as an activation-specific probe for Ras.
Oncogene (1997-02-06)
J de Rooij, J L Bos
ABSTRACT
Ras is a small GTPase that cycles between an inactive GDP-bound and an active GTP-bound form. A large variety of ligands that stimulate cell surface receptors induce the activation of Ras. Thus far, this activation could only be measured by the increase of GTP bound to Ras, which was precipitated from radio-labelled cell extract. We have used the minimal Ras-binding domain (RBD) of Raf1 (aa 51-131) to identify in vivo activated Ras. This novel method is based on the observation that RBD binds RasGTP in vitro with a Kd of 20 nM whereas the affinity between RBD and RasGDP is three orders of magnitude lower. Here we show that the Gst-RBD fusion protein precipitates transfected RasL61 (RasGTP) but not RasN17 (RasGDP) from cell lysates. In addition, we demonstrate for two different cell lines that the increase in RasGTP is reflected by an increase in Ras bound to Gst-RBD. From these results we conclude that the minimal Ras-binding domain of Raf1 is an excellent activation specific-probe for Ras.
Sigma-Aldrich
Raf-1 RBD Protein, GST, 300 µg, GST fusion-protein, corresponding to the human Ras Binding Domain (RBD, residues 1-149) of Raf-1, expressed in E. coli. with purity 50% at full length molecular weight 42 kDa. For use in Affinity Binding Assays
Sigma-Aldrich
Ras Assay Reagent (Raf-1 RBD, agarose)
Sigma-Aldrich
Ras Activation Assay Kit
Sigma-Aldrich
Ral Activation Assay Kit, Non-radioactive Ral Activation Assay Kit can be used to precipitate Ral-A-GTP from cell lysates & detection by a Ral-A specific antibody.