Merck
  • Synthesis and topoisomerase I inhibitory activity of a novel diazaindeno[2,1-b]phenanthrene analogue of Lamellarin D.
Synthesis and topoisomerase I inhibitory activity of a novel diazaindeno[2,1-b]phenanthrene analogue of Lamellarin D.
Bioorganic & medicinal chemistry (2011-07-26)
Salvatore Cananzi, Lucio Merlini, Roberto Artali, Giovanni Luca Beretta, Nadia Zaffaroni, Sabrina Dallavalle
ABSTRACT
A novel 5-oxa-6a,8-diazaindeno[2,1-b]phenanthren-7-one scaffold was designed and synthesized as an active analogue of the cytotoxic marine alkaloid Lamellarin D. The design was based on molecular modeling of the site of interaction of Lamellarin D with DNA-topoisomerase I cleavable complex, whereas the synthesis capitalized on a simple Friedel-Crafts cyclization of indole to a β-carbolinone nucleus. The product exhibited topoisomerase I poisoning activity and submicromolar cytotoxicity on human non-small cell lung cancer H460 cell line.
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis