Merck
  • Acetyl CoA Carboxylase Inhibition Reduces Hepatic Steatosis but Elevates Plasma Triglycerides in Mice and Humans: A Bedside to Bench Investigation.
Acetyl CoA Carboxylase Inhibition Reduces Hepatic Steatosis but Elevates Plasma Triglycerides in Mice and Humans: A Bedside to Bench Investigation.
Cell metabolism (2017-08-03)
Chai-Wan Kim, Carol Addy, Jun Kusunoki, Norma N Anderson, Stanislaw Deja, Xiaorong Fu, Shawn C Burgess, Cai Li, Marcie Ruddy, Manu Chakravarthy, Steve Previs, Stuart Milstein, Kevin Fitzgerald, David E Kelley, Jay D Horton
ABSTRACT
Inhibiting lipogenesis prevents hepatic steatosis in rodents with insulin resistance. To determine if reducing lipogenesis functions similarly in humans, we developed MK-4074, a liver-specific inhibitor of acetyl-CoA carboxylase (ACC1) and (ACC2), enzymes that produce malonyl-CoA for fatty acid synthesis. MK-4074 administered to subjects with hepatic steatosis for 1 month lowered lipogenesis, increased ketones, and reduced liver triglycerides by 36%. Unexpectedly, MK-4074 increased plasma triglycerides by 200%. To further investigate, mice that lack ACC1 and ACC2 in hepatocytes (ACC dLKO) were generated. Deletion of ACCs decreased polyunsaturated fatty acid (PUFA) concentrations in liver due to reduced malonyl-CoA, which is required for elongation of essential fatty acids. PUFA deficiency induced SREBP-1c, which increased GPAT1 expression and VLDL secretion. PUFA supplementation or siRNA-mediated knockdown of GPAT1 normalized plasma triglycerides. Thus, inhibiting lipogenesis in humans reduced hepatic steatosis, but inhibiting ACC resulted in hypertriglyceridemia due to activation of SREBP-1c and increased VLDL secretion.
Sigma-Aldrich
Pioglitazone hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-CPT1A Antibody, serum, from rabbit