Merck
  • Adhesion characteristics and stability assessment of lectin-modified liposomes for site-specific drug delivery.
Adhesion characteristics and stability assessment of lectin-modified liposomes for site-specific drug delivery.
Biochimica et biophysica acta (2007-10-30)
Heike Bakowsky, Thomas Richter, Carsten Kneuer, Dick Hoekstra, Ulrich Rothe, Gerd Bendas, Carsten Ehrhardt, Udo Bakowsky
ABSTRACT
Carbohydrate moieties of the cellular glycocalyx have been suggested to play an important role in biological recognition processes during pathologic conditions, such as inflammation and cancer. Herein, we describe lectin-modified liposomes which might have potential for site-specific drug delivery during the therapy of such diseases. Specific interactions of plain (i.e., unmodified) and PEGylated, lectin-grafted liposomes with model membranes were investigated under real-time flow conditions using a quartz crystal microbalance. In addition, the morphology of the liposomal systems was assessed by atomic force microscopy. Plain liposomes exhibited only unspecific adhesion to glycolipid membranes and had a tendency to coalesce. The degree of membrane interaction was significantly increased when plain liposomes were modified with the lectin, Concanavalin A. However, vesicle fusion also markedly increased as a result of lectin modification. Additional PEGylation of liposomes reduced unspecific adhesion phenomena, as well as coalescence. Moreover, our studies enabled us to establish quartz crystal microbalance and atomic force microscopy as powerful and complementary methods to characterize adhesion properties of targeted drug delivery systems.
Product Description
Avanti
DSPE-PEG(2000) Cyanur, Avanti Research - A Croda Brand 880122P, powder
Avanti
16:0 Cyanur PE, Avanti Research - A Croda Brand 870287P, powder