Merck
  • The Arabidopsis LecRK-VI.2 associates with the pattern-recognition receptor FLS2 and primes Nicotiana benthamiana pattern-triggered immunity.
The Arabidopsis LecRK-VI.2 associates with the pattern-recognition receptor FLS2 and primes Nicotiana benthamiana pattern-triggered immunity.
The Plant journal : for cell and molecular biology (2014-05-23)
Pin-Yao Huang, Yu-Hung Yeh, An-Chi Liu, Chiu-Ping Cheng, Laurent Zimmerli
ABSTRACT
Pattern-triggered immunity (PTI) is broad spectrum and manipulation of PTI is believed to represent an attractive way to engineer plants with broad-spectrum disease resistance. PTI is activated upon perception of microbe-associated molecular patterns (MAMPs) by pattern-recognition receptors (PRRs). We have recently demonstrated that the L-type lectin receptor kinase-VI.2 (LecRK-VI.2) positively regulates Arabidopsis thaliana PTI. Here we show through in vitro pull-down, bimolecular fluorescence complementation and co-immunoprecipitation analyses that LecRK-VI.2 associates with the PRR FLS2. We also demonstrated that LecRK-VI.2 from the cruciferous plant Arabidopsis remains functional after interfamily transfer to the Solanaceous plant Nicotiana benthamiana. Wild tobacco plants ectopically expressing LecRK-VI.2 were indeed more resistant to virulent hemi-biotrophic and necrotrophic bacteria, but not to the fungal pathogen Botrytis cinerea suggesting that, as with Arabidopsis, the LecRK-VI.2 protective effect in N. benthamiana is bacteria specific. Ectopic expression of LecRK-VI.2 in N. benthamiana primed PTI-mediated reactive oxygen species production, mitogen-activated protein kinase (MAPK) activity, callose deposition and gene expression upon treatment with the MAMP flagellin. Our findings identified LecRK-VI.2 as a member of the FLS2 receptor complex and suggest that heterologous expression of components of PRR complexes can be used as tools to engineer plant disease resistance to bacteria.
Sigma-Aldrich
Monoclonal Anti-Maltose Binding Protein antibody produced in mouse, clone MBP-17, ascites fluid