- Feedback regulation of the heat shock response.
Feedback regulation of the heat shock response.
Handbook of experimental pharmacology (2006-04-14)
R Voellmy
PMID16610354
ABSTRACT
The heat shock response is triggered primarily by nonnative proteins accumulating in a stressed cell and results in increased expression of heat shock proteins (Hsps), i.e., of chaperones capable of participating in the refolding or elimination of nonnative proteins. Best known is the transcriptional part of this response that is mediated predominantly by heat shock factor 1 (HSF1). HSF1 activity is regulated at different levels by Hsps and co-chaperones and is modulated further by a number of mechanisms involving other stress-regulated aspects of cell metabolism.
Sigma-Aldrich
HSF1, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution