Merck
  • Bad, a heterodimeric partner for Bcl-XL and Bcl-2, displaces Bax and promotes cell death.
Bad, a heterodimeric partner for Bcl-XL and Bcl-2, displaces Bax and promotes cell death.
Cell (1995-01-27)
E Yang, J Zha, J Jockel, L H Boise, C B Thompson, S J Korsmeyer
ABSTRACT
To extend the mammalian cell death pathway, we screened for further Bcl-2 interacting proteins. Both yeast two-hybrid screening and lambda expression cloning identified a novel interacting protein, Bad, whose homology to Bcl-2 is limited to the BH1 and BH2 domains. Bad selectively dimerized with Bcl-xL as well as Bcl-2, but not with Bax, Bcl-xs, Mcl-1, A1, or itself. Bad binds more strongly to Bcl-xL than Bcl-2 in mammalian cells, and it reversed the death repressor activity of Bcl-xL, but not that of Bcl-2. When Bad dimerized with Bcl-xL, Bax was displaced and apoptosis was restored. When approximately half of Bax was heterodimerized, death was inhibited. The susceptibility of a cell to a death signal is determined by these competing dimerizations in which levels of Bad influence the effectiveness of Bcl-2 versus Bcl-xL in repressing death.
Sigma-Aldrich
BAD, GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution