Merck
Inhibition of JNK activation through NF-kappaB target genes.
Nature (2001-11-20)
G Tang, Y Minemoto, B Dibling, N H Purcell, Z Li, M Karin, A Lin
ABSTRACT
The proinflammatory cytokine tumour necrosis factor-alpha (TNF-alpha) regulates immune responses, inflammation and programmed cell death (apoptosis). The ultimate fate of a cell exposed to TNF-alpha is determined by signal integration between its different effectors, including IkappaB kinase (IKK), c-Jun N-terminal protein kinase (JNK) and caspases. Activation of caspases is required for apoptotic cell death, whereas IKK activation inhibits apoptosis through the transcription factor NF-kappaB, whose target genes include caspase inhibitors. JNK activates the transcription factor c-Jun/AP-1, as well as other targets. However, the role of JNK activation in apoptosis induced by TNF-alpha is less clear. It is unknown whether any crosstalk occurs between IKK and JNK, and, if so, how it affects TNF-alpha-induced apoptosis. We investigated this using murine embryonic fibroblasts that are deficient in either the IKKbeta catalytic subunit of the IKK complex or the RelA/p65 subunit of NF-kappaB. Here we show that in addition to inhibiting caspases, the IKK/NF-kappaB pathway negatively modulates TNF-alpha-mediated JNK activation, partly through NF-kappaB-induced X-chromosome-linked inhibitor of apoptosis (XIAP). This negative crosstalk, which is specific to TNF-alpha signalling and does not affect JNK activation by interleukin-1 (IL-1), contributes to inhibition of apoptosis.
Sigma-Aldrich
XIAP, GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution