Merck
  • Phosphorylation of androgen receptor serine 81 is associated with its reactivation in castration-resistant prostate cancer.
Phosphorylation of androgen receptor serine 81 is associated with its reactivation in castration-resistant prostate cancer.
Cancer letters (2018-09-16)
Joshua W Russo, Xiaming Liu, Huihui Ye, Carla Calagua, Sen Chen, Olga Voznesensky, James Condulis, Fen Ma, Mary-Ellen Taplin, David J Einstein, Steven P Balk, Shaoyong Chen
ABSTRACT
Phosphorylation of serine 81 (pS81) in the N-terminal transactivation domain of the androgen receptor (AR) has been linked to its transcriptional activation in prostate cancer (PCa) cell lines, but in vivo studies have been limited. Moreover, the role of pS81 in the reactivation of AR when tumors relapse after androgen deprivation therapy (castration-resistant prostate cancer, CRPC) has not been determined. In this study we validate a pS81 antibody for immunohistochemistry (IHC) and show it yields strong nuclear staining in primary PCa clinical samples and in the VCaP PCa xenograft model. Moreover, this staining was decreased at 7 days post-castration in VCaP xenografts, coinciding with markedly decreased AR transcriptional activity. Staining with the pS81 antibody then was restored when the VCaP xenografts relapsed, which was associated with restoration of AR transcriptional activity. Significantly, analysis of CRPC clinical samples, including tumors that had progressed during treatment with abiraterone, showed strong nuclear staining with the pS81 antibody. Together these findings indicate that AR reactivation in CRPC is associated with S81 phosphorylation, and suggest that IHC for pS81 may be useful as a biomarker of AR activity in CRPC.
Sigma-Aldrich
Mayer′s Hematoxylin Solution
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Anti-phospho-Androgen Receptor (Ser81) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Anti-Tubulin Antibody, beta, clone KMX-1, clone KMX-1, Chemicon®, from mouse
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade