Merck
  • Effect of yeast volatile organic compounds on ochratoxin A-producing Aspergillus carbonarius and A. ochraceus.
Effect of yeast volatile organic compounds on ochratoxin A-producing Aspergillus carbonarius and A. ochraceus.
International journal of food microbiology (2018-07-11)
Maria Grazia Farbo, Pietro Paolo Urgeghe, Stefano Fiori, Angela Marcello, Stefania Oggiano, Virgilio Balmas, Zahoor Ul Hassan, Samir Jaoua, Quirico Migheli
ABSTRACT
Many foods and beverages in temperate and tropical regions are prone to contamination by ochratoxin A (OTA), one of the most harmful mycotoxins for human and animal health. Aspergillus ochraceus and Aspergillus carbonarius are considered among the main responsible for OTA contamination. We have previously demonstrated that four low or non- fermenting yeasts are able to control the growth and sporulation of OTA-producing Aspergilli both in vitro and on detached grape berries: the biocontrol effect was partly due to the release of volatile organic compounds (VOCs). Aiming to further characterise the effect of VOCs produced by biocontrol yeast strains, we observed that, beside vegetative growth and sporulation, the volatile compounds significantly reduced the production of OTA by two A. carbonarius and A. ochraceus isolates. Exposure to yeast VOCs also affected gene expression in both species, as confirmed by downregulation of polyketide synthase, non-ribosomal peptide synthase, monooxygenase, and the regulatory genes laeA and veA. The main compound of yeast VOCs was 2-phenylethanol, as detected by Headspace-Solid Phase Microextraction-Gas Chromatography-Tandem Mass Spectrometry (HS-SPME-GC-MS) analysis. Yeast VOCs represent a promising tool for the containment of growth and development of mycotoxigenic fungi, and a valuable aid to guarantee food safety and quality.
Product Description
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, matrix active group 50/30µm DVB/CAR/PDMS, needle size 24 ga, StableFlex fiber, for use with autosampler, fiber L 2 cm, pk of 3
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly, for use with autosampler, matrix active group 50/30µm DVB/CAR/PDMS, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder