- Transcriptomic response to GABA-producing Lactobacillus plantarum CGMCC 1.2437T induced by L-MSG.
Transcriptomic response to GABA-producing Lactobacillus plantarum CGMCC 1.2437T induced by L-MSG.
Gamma-aminobutyric acid (GABA) is an inhibitory neurotransmitter found in the central nervous system of mammals. A range of bacterial species can synthesize GABA, including Lactobacillus plantarum of which L-monosodium glutamate (L-MSG) is an inducer of its production. In order to synthesize GABA in high concentrations, L-MSG was utilized as the single inducing factor, a chemically defined medium (CDM) was used as the fermentation substrate, with L. plantarum CGMCC 1.2437T cultured in medium supplemented with or without L-MSG. High-throughput transcriptome sequencing was used to explore the differential genes expression of bacterial cells at 36 h of fermentation, where the GABA concentration of CDM with L-MSG reached the peak value and was 7.7 times higher than that of medium without L-MSG at the same timepoint. A total of 87 genes showed significant differential expression induced by L-MSG: of these, 69 were up-regulated genes and 18 were down-regulated. The up-regulated genes were assigned to biological processes and molecular function, while the down-regulated genes covered biological process, cellular process and molecular function. Interrogation of results using Gene Ontology (GO) and Kyoto Encyclopedia of Genes and Genomes (KEGG) analyses, indicated carbohydrate metabolism, fatty acid synthesis and amino acid metabolism were closely associated with GABA synthesis induced by L-MSG. This study provides insights into L. plantarum-mediated GABA fermentation at the molecular level and will provide a new approach for further studies related to GABA production by the other Lactic acid bacteria.