Merck
  • Epigenetic Therapy Ties MYC Depletion to Reversing Immune Evasion and Treating Lung Cancer.
Epigenetic Therapy Ties MYC Depletion to Reversing Immune Evasion and Treating Lung Cancer.
Cell (2017-12-02)
Michael J Topper, Michelle Vaz, Katherine B Chiappinelli, Christina E DeStefano Shields, Noushin Niknafs, Ray-Whay Chiu Yen, Alyssa Wenzel, Jessica Hicks, Matthew Ballew, Meredith Stone, Phuoc T Tran, Cynthia A Zahnow, Matthew D Hellmann, Valsamo Anagnostou, Pamela L Strissel, Reiner Strick, Victor E Velculescu, Stephen B Baylin
ABSTRACT
Combining DNA-demethylating agents (DNA methyltransferase inhibitors [DNMTis]) with histone deacetylase inhibitors (HDACis) holds promise for enhancing cancer immune therapy. Herein, pharmacologic and isoform specificity of HDACis are investigated to guide their addition to a DNMTi, thus devising a new, low-dose, sequential regimen that imparts a robust anti-tumor effect for non-small-cell lung cancer (NSCLC). Using in-vitro-treated NSCLC cell lines, we elucidate an interferon α/β-based transcriptional program with accompanying upregulation of antigen presentation machinery, mediated in part through double-stranded RNA (dsRNA) induction. This is accompanied by suppression of MYC signaling and an increase in the T cell chemoattractant CCL5. Use of this combination treatment schema in mouse models of NSCLC reverses tumor immune evasion and modulates T cell exhaustion state towards memory and effector T cell phenotypes. Key correlative science metrics emerge for an upcoming clinical trial, testing enhancement of immune checkpoint therapy for NSCLC.
Sigma-Aldrich
Anti-DNMT1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
BrdU Cell Proliferation Assay, This proliferation assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells.