Merck
Neurons generated from carcinoma stem cells support cancer progression.
Signal transduction and targeted therapy (2017-12-22)
Ran Lu, Chuanwen Fan, Wenqi Shangguan, Yuan Liu, Yu Li, Yanna Shang, Dongqin Yin, Shengliang Zhang, Qiaorong Huang, Xue Li, Wentong Meng, Hong Xu, Zongguang Zhou, Jiankun Hu, Weimin Li, Lunxu Liu, Xianming Mo
ABSTRACT
Recent evidences show that nervous system acts as a crucial part of cancer microenvironment. Infiltration of nerve fibers into cancer microenvironment has an important active role in cancer progression. The stimulations of both cancer growth and metastasis by members of nervous system such as neurons and glial cells have been demonstrated. However, how the nervous system is built in cancer is largely unknown. Here we show that a fraction of cancer stem cells (CSCs) derived from patients with gastric carcinoma and colorectal carcinoma are capable of producing neurons that are involved in tumor neurogenesis and tumor growth. Cancer stem cell monoclone derived from a single cancer stem cell was able to generate neurons including sympathetic and parasympathetic neurons to take part in the nervous system in cancer tissues. Knocking down the neural cell generating capability of the human CSCs inhibited the growth of xenograft tumors in mouse model. Our data demonstrate that human CSCs are able to produce one of most important components in the cancer microenvironment that are required for cancer development and progression.
Sigma-Aldrich
Anti-VAChT (C-terminal) antibody produced in rabbit, IgG fraction of antiserum