Merck
Visible-Light-Promoted Synthesis of Dibenzofuran Derivatives.
The Journal of organic chemistry (2017-12-14)
Ji Young Cho, Geum-Bee Roh, Eun Jin Cho
ABSTRACT
Dibenzofurans are naturally occurring molecules that have received considerable attention for a variety of practical applications, such as in pharmaceuticals and electronic materials. Herein, an efficient and eco-friendly method for the synthesis of dibenzofuran derivatives via intramolecular C-O bond formation, which involves the in situ production of a diazonium salt, is described. The transformation requires a diazotizing agent and is promoted by the use of an organic photosensitizer under visible-light irradiation.
Sigma-Aldrich
2,4,6-Tri-(4-fluorophenyl)pyrylium tetrafluoroborate, ≥95%