
Zolpidem for system suitability

Synonym(s):
Zolpidem, N,N,6-Trimethyl-2-(4-methylphenyl)-imidazo[1,2-a]pyridine-3-acetamide
Empirical Formula (Hill Notation):
C19H21N3O
CAS Number:
Molecular Weight:
307.39
Products
Formula Weight
Gene Alias
Manufacturer
Quality Segment
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing