![Zinc trifluoroacetate hydrate](/deepweb/assets/sigmaaldrich/product/structures/311/787/452e60de-ee0a-49e8-98c9-ad4a716f1606/640/452e60de-ee0a-49e8-98c9-ad4a716f1606.png)
Zinc trifluoroacetate hydrate
![Zinc trifluoroacetate hydrate](/deepweb/assets/sigmaaldrich/product/structures/311/787/452e60de-ee0a-49e8-98c9-ad4a716f1606/640/452e60de-ee0a-49e8-98c9-ad4a716f1606.png)
Synonym(s):
Trifluoroacetic acid zinc salt
Linear Formula:
Zn(CF3COO)2 · xH2O
CAS Number:
Molecular Weight:
291.42 (anhydrous basis)
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing