![Zilpaterol hydrochloride VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/204/274/d755d4a4-7e66-43a9-b3d9-205437f6ab2d/640/d755d4a4-7e66-43a9-b3d9-205437f6ab2d.png)
Zilpaterol hydrochloride
![Zilpaterol hydrochloride VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/204/274/d755d4a4-7e66-43a9-b3d9-205437f6ab2d/640/d755d4a4-7e66-43a9-b3d9-205437f6ab2d.png)
Synonym(s):
(6RS,7RS)-4,5,6, 7-Tetrahydro-7-hydroxy-6-[(1-methylethyl)amino]imidazo[ 4,5,1-jk][1]benzazepin-2(1H)-one hydrochloride
Empirical Formula (Hill Notation):
C14H19N3O2 · HCl
CAS Number:
Molecular Weight:
297.78
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing