Zanamivir for system suitability
Synonym(s):
Zanamivir, 4-Guanidino-2,4-dideoxy-2,3-dehydro-N-acetylneuraminic acid, 4-Guanidino-Neu5Ac2en
Empirical Formula (Hill Notation):
C12H20N4O7
CAS Number:
Molecular Weight:
332.31
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing