Yangonin
Synonym(s):
4-Methoxy-6-((E)-4-methoxystyryl)pyran-2-one
Empirical Formula (Hill Notation):
C15H14O4
CAS Number:
Molecular Weight:
258.27
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing