
Voriconazole impurity E

Synonym(s):
(±)-Camphor-10-sulfonic acid, (±)-β-Camphorsulfonic acid
Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing