![Verapamil Impurity M](/deepweb/assets/sigmaaldrich/product/images/294/481/30963d1d-9cb2-4457-b6b5-e0fc913555dd/640/30963d1d-9cb2-4457-b6b5-e0fc913555dd.jpg)
Verapamil Impurity M
![Verapamil Impurity M](/deepweb/assets/sigmaaldrich/product/images/294/481/30963d1d-9cb2-4457-b6b5-e0fc913555dd/640/30963d1d-9cb2-4457-b6b5-e0fc913555dd.jpg)
Synonym(s):
5,5′-[[2-(3,4-Dimethoxyphenyl)ethyl]imino]bis[2-(3,4-dimethoxyphenyl)-2-(1-methylethyl)pentanenitrile] hydrochloride
Empirical Formula (Hill Notation):
C42H57N3O6 · HCl
CAS Number:
Molecular Weight:
736.38
Products
Brand
Formula Weight
Physical Form
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing