![TSKgel® ODS-120A Guard Cartridge phase C18 (octadecyl), L × I.D. 1.5 cm × 3.2 mm, 5 μm particle size, pkg of 3 ea](/deepweb/assets/sigmaaldrich/product/images/383/940/01283a0b-f7a9-4b36-9d35-92fb268696d9/640/01283a0b-f7a9-4b36-9d35-92fb268696d9.jpg)
TSKgel® ODS-120A (5 µm) HPLC Columns
![TSKgel® ODS-120A Guard Cartridge phase C18 (octadecyl), L × I.D. 1.5 cm × 3.2 mm, 5 μm particle size, pkg of 3 ea](/deepweb/assets/sigmaaldrich/product/images/383/940/01283a0b-f7a9-4b36-9d35-92fb268696d9/640/01283a0b-f7a9-4b36-9d35-92fb268696d9.jpg)
Synonym(s):
TSKgel® Reversed Phase HPLC Column, TSKgel® Reversed Phase HPLC Guard Cartridge
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing