![TSKgel® G-Oligo-PW Guard Column phase hydroxylated methacrylate, L × I.D. 4 cm × 6 mm, 13 μm particle size](/deepweb/assets/sigmaaldrich/product/images/760/943/644baf15-a9ae-4104-9fa4-fecb1d907903/640/644baf15-a9ae-4104-9fa4-fecb1d907903.jpg)
TSKgel® G-Oligo-PW (13 µm) HPLC Columns
![TSKgel® G-Oligo-PW Guard Column phase hydroxylated methacrylate, L × I.D. 4 cm × 6 mm, 13 μm particle size](/deepweb/assets/sigmaaldrich/product/images/760/943/644baf15-a9ae-4104-9fa4-fecb1d907903/640/644baf15-a9ae-4104-9fa4-fecb1d907903.jpg)
Synonym(s):
TSKgel® Size Exclusion (PW-Type) HPLC Guard Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing