Tris(diethylamido)(tert-butylimido)niobium(V)
Synonym(s):
t-Butylimido tris(diethylamido)niobium(V)
Linear Formula:
((CH3)6N)3NbN(CH3)3
CAS Number:
Molecular Weight:
380.41
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing