Skip to Content
Merck
Triclocarban Related Compound C pharmaceutical secondary standard, certified reference material

Triclocarban Related Compound C

Triclocarban Related Compound C pharmaceutical secondary standard, certified reference material
Synonym(s):
1,3-Bis(3,4-dichlorophenyl)urea, 3,3′,4,4′-Tetrachlorocarbanilide
Empirical Formula (Hill Notation):
C13H8Cl4N2O
CAS Number:
Molecular Weight:
350.03

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2