![Tranexamic acid impurity standard British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/129/772/2af533ab-ebc9-45b6-a5ee-330ea2798427/640/2af533ab-ebc9-45b6-a5ee-330ea2798427.png)
Tranexamic acid impurity standard
![Tranexamic acid impurity standard British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/129/772/2af533ab-ebc9-45b6-a5ee-330ea2798427/640/2af533ab-ebc9-45b6-a5ee-330ea2798427.png)
Synonym(s):
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, AMCA, AMCHA, HAKU, TAMCHA, Tranexamic acid
Linear Formula:
H2NCH2C6H10CO2H
CAS Number:
Molecular Weight:
157.21
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing