Tiotropium Related Compound C
Synonym(s):
(1R,3S,5S)-3-[2-Hydroxy-2,2-di(thiophen-2-yl)acetoxy]-8,8-dimethyl-8- azabicyclo[3.2.1]oct-6-en-8-ium bromide, (3-endo)- 3-[(Hydroxydi-2-thienylacetyl)oxy]-8,8-dimethyl-8-azoniabicyclo[3.2.1]oct-6-ene bromide
Empirical Formula (Hill Notation):
C19H22BrNO3S2
CAS Number:
Molecular Weight:
456.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing