Tetrandrine
Synonym(s):
(1β)-6,6′,7,12-Tetramethoxy-2,2′-dimethylberbaman
Empirical Formula (Hill Notation):
C38H42N2O6
CAS Number:
Molecular Weight:
622.75
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4