![Terpin monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/342/328/4d560f06-9b65-4c9a-b808-b6e9909d7589/640/4d560f06-9b65-4c9a-b808-b6e9909d7589.png)
Terpin monohydrate
![Terpin monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/342/328/4d560f06-9b65-4c9a-b808-b6e9909d7589/640/4d560f06-9b65-4c9a-b808-b6e9909d7589.png)
Synonym(s):
p-Menthane-1,8-diol monohydrate, cis-4-Hydroxy-α,α,4-trimethylcyclohexanemethanol monohydrate, cis-Terpin monohydrate, cis-p-Menthane-1,8-diol monohydrate
Empirical Formula (Hill Notation):
C10H20O2 · H2O
CAS Number:
Molecular Weight:
190.28
Products
Formula Weight
Manufacturer
Format
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing