Skip to Content
Merck
Syringaldazine 98%

Syringaldazine

Syringaldazine 98%
Synonym(s):
4-Hydroxy-3,5-dimethoxybenzaldehyde azine
Linear Formula:
[HOC6H2(OCH3)2CH=N-]2
CAS Number:
Molecular Weight:
360.36
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment