![Sulfadimidine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/145/752/5fb6d2b6-664c-425c-8487-8317fed07cd1/640/5fb6d2b6-664c-425c-8487-8317fed07cd1.png)
Sulfadimidine
![Sulfadimidine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/145/752/5fb6d2b6-664c-425c-8487-8317fed07cd1/640/5fb6d2b6-664c-425c-8487-8317fed07cd1.png)
Synonym(s):
Sulfamethazine, 4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine
Empirical Formula (Hill Notation):
C12H14N4O2S
CAS Number:
Molecular Weight:
278.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing