Skip to Content
Merck
(S)-(−)-2-Acetoxypropionyl chloride 97%

(S)-(−)-2-Acetoxypropionyl chloride

(S)-(−)-2-Acetoxypropionyl chloride 97%
Synonym(s):
O-Acetyl-L-lactyl chloride, AP-Cl
Linear Formula:
CH3CH(O2CCH3)COCl
CAS Number:
Molecular Weight:
150.56
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing