![Ropinirole for peak identification 3 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/414/322/cd291e08-e92b-449b-9084-120a3f20f0ee/640/cd291e08-e92b-449b-9084-120a3f20f0ee.png)
Ropinirole for peak identification 3
![Ropinirole for peak identification 3 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/414/322/cd291e08-e92b-449b-9084-120a3f20f0ee/640/cd291e08-e92b-449b-9084-120a3f20f0ee.png)
Synonym(s):
Ropinirole hydrochloride, 4-[2-(Dipropylamino)ethyl]-1,3-dihydro-2H-indol-2-one hydrochloride, SKF 101468 hydrochloride
Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84
Products
Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing