Skip to Content
Merck
R(+)-HA-966 solid

R(+)-HA-966

R(+)-HA-966 solid
Synonym(s):
R(+)-3-Amino-1-hydroxy-2-pyrrolidinone
Empirical Formula (Hill Notation):
C4H8N2O2
CAS Number:
Molecular Weight:
116.12

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing