![Quercetin 3-glucoside analytical standard](/deepweb/assets/sigmaaldrich/product/structures/385/032/bf00357c-3ee2-4ac0-b82d-97e1ce0b42af/640/bf00357c-3ee2-4ac0-b82d-97e1ce0b42af.png)
Quercetin 3-glucoside
![Quercetin 3-glucoside analytical standard](/deepweb/assets/sigmaaldrich/product/structures/385/032/bf00357c-3ee2-4ac0-b82d-97e1ce0b42af/640/bf00357c-3ee2-4ac0-b82d-97e1ce0b42af.png)
Synonym(s):
Quercetin 3-β-D-glucoside, 3,3′,4′,5,7-Pentahydroxyflavone 3-β-glucoside, Isoquercitrin
Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing