Podophyllotoxin
Empirical Formula (Hill Notation):
C22H22O8
CAS Number:
Molecular Weight:
414.41
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing