Skip to Content
Merck
Pirbuterol acetate VETRANAL®, analytical standard

Pirbuterol acetate

Pirbuterol acetate VETRANAL®, analytical standard
Synonym(s):
(RS)-2-(tert-Butylamino)-1-(6-hydroxymethyl-5-hydroxy-2-pyridyl)ethanol acetate salt, 6-(2-tert-Butylamino-1-hydroxyethyl)-2-hydroxymethylpyridin-3-ol acetate
Empirical Formula (Hill Notation):
C12H20N2O3 · C2H4O2
CAS Number:
Molecular Weight:
300.35
EC Number:

Products

Brand
Formula Weight
Gene Alias
Product Line
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing