
Phosphatidylcholine (Soy)

Synonym(s):
Phosphatidylcholine (Soy)
Empirical Formula (Hill Notation):
C42H80NO8P
CAS Number:
Molecular Weight:
758.06
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing