Skip to Content
Merck
vibrant-m

Phosphatidylcholine from soya bean

vibrant-m
Synonym(s):
Phosphatidylcholine (Soy)
Empirical Formula (Hill Notation):
C42H80NO8P
CAS Number:
Molecular Weight:
758.06
EC Number:

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1