![Phosphatase, Alkaline shrimp ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host](/deepweb/assets/sigmaaldrich/product/images/333/043/d5d47c09-141e-4359-ae14-4e490e17affa/640/d5d47c09-141e-4359-ae14-4e490e17affa.jpg)
Phosphatase, Alkaline shrimp
![Phosphatase, Alkaline shrimp ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host](/deepweb/assets/sigmaaldrich/product/images/333/043/d5d47c09-141e-4359-ae14-4e490e17affa/640/d5d47c09-141e-4359-ae14-4e490e17affa.jpg)
Synonym(s):
Phosphatase, Alkaline from shrimp, Orthophosphoric-monoester phosphohydrolase (alkaline optimum)
CAS Number:
Products
Brand
Physical Form
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing