![Phenytoin Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/427/190/4be6a4b2-014e-4764-b516-1cbff6655433/640/4be6a4b2-014e-4764-b516-1cbff6655433.png)
Phenytoin Related Compound A
![Phenytoin Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/427/190/4be6a4b2-014e-4764-b516-1cbff6655433/640/4be6a4b2-014e-4764-b516-1cbff6655433.png)
Synonym(s):
2,2-Diphenylglycine
Linear Formula:
H2NC(C6H5)2CO2H
CAS Number:
Molecular Weight:
227.26
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing