Skip to Content
Merck
p-Cymene phyproof® Reference Substance

p-Cymene

p-Cymene phyproof® Reference Substance
Synonym(s):
1-Isopropyl-4-methylbenzene, 4-Isopropyltoluene
Linear Formula:
CH3C6H4CH(CH3)2
CAS Number:
Molecular Weight:
134.22

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing