![O-Desmethylnaproxen analytical standard](/deepweb/assets/sigmaaldrich/product/structures/412/719/e11eb698-2d5e-44cf-9e1f-416c635dfffd/640/e11eb698-2d5e-44cf-9e1f-416c635dfffd.png)
O-Desmethylnaproxen
![O-Desmethylnaproxen analytical standard](/deepweb/assets/sigmaaldrich/product/structures/412/719/e11eb698-2d5e-44cf-9e1f-416c635dfffd/640/e11eb698-2d5e-44cf-9e1f-416c635dfffd.png)
Synonym(s):
(S)-6-Hydroxy-α-methyl-2-naphthaleneacetic acid, d-2-(6-Hydroxy-2-naphthyl)proprionic acid
Empirical Formula (Hill Notation):
C13H12O3
CAS Number:
Molecular Weight:
216.23
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing