Noroxymorphone hydrochloride
Synonym(s):
(5α)-4,5-Epoxy-3,14-dihydroxymorphinan-6-one monohydrochloride, Noroxymorphone hydrochloride
Empirical Formula (Hill Notation):
C16H17NO4 · HCl
CAS Number:
Molecular Weight:
323.77
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2