![Nonadecane analytical standard](/deepweb/assets/sigmaaldrich/product/structures/878/317/866d87e5-a554-48fe-85c5-3a0d8f80011a/640/866d87e5-a554-48fe-85c5-3a0d8f80011a.png)
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing