Skip to Content
Merck
Nicotinamide Impurity B Pharmaceutical Secondary Standard; Certified Reference Material

Nicotinamide Impurity B

Nicotinamide Impurity B Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3-Pyridinecarbonitrile, 3-Cyanopyridine, Nicotinic acid nitrile, Nicotinonitrile
Empirical Formula (Hill Notation):
C6H4N2
CAS Number:
Molecular Weight:
104.11

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1