Skip to Content
Merck
Nickel(II) sulfate hexahydrate ≥99.99% trace metals basis

Nickel(II) sulfate hexahydrate

Nickel(II) sulfate hexahydrate ≥99.99% trace metals basis
Synonym(s):
Nickel(II) sulfate hexahydrate, Nickelous Sulfate, Hexahydrate
Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85

Products

Product Category
Brand
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade