Skip to Content
Merck
n-Dodecyl β-D-maltoside BioXtra, ≥98% (GC)

n-Dodecyl β-D-maltoside

n-Dodecyl β-D-maltoside BioXtra, ≥98% (GC)
Synonym(s):
DDM, Lauryl-β-D-maltoside
Empirical Formula (Hill Notation):
C24H46O11
CAS Number:
Molecular Weight:
510.62

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing