
Narciclasine

Synonym(s):
Narciclasine, (2S-(2-alpha,3-beta,4-beta,4a-beta))-3,4,4a,5-tetrahydro-2,3,4,7-tetrahydroxy-(1,3)Dioxolo(4,5-j)phenanthridin-6(2H)-one, 3,4,4a,5-Tetrahydro-2,3,4,7-tetrahydroxy-(1,3)dioxolo(4,5-j)phenanthridin-6(2H)-one, BRN 1087400, Lycoricidin-A, Lycoricidinol, NSC 266535
Empirical Formula (Hill Notation):
C14H13NO7
CAS Number:
Molecular Weight:
307.26
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing